Invitrogen
Manufacturer Code:PA527240
Catalog # PIPA527240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 75 and 154 of Human SOD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALS ALS1 IPOA SOD hSod1 homodimer Cu/Zn superoxide dismutase SOD soluble indophenoloxidase A superoxide dismutase &lsqbCu-Zn] superoxide dismutase cystolic; Cu(2+)-Zn2+ superoxide dismutase; Cu/Zn superoxide dismutase; epididymis secretory protein Li 44; hSod1; indophenoloxidase A; SOD, soluble; Superoxide dismutase 1; Superoxide dismutase [Cu-Zn]; superoxide dismutase, cystolic
Gene Aliases: ALS; ALS1; B430204E11Rik; Cu/Zn-SOD; CuZnSOD; HEL-S-44; homodimer; hSod1; Ipo-1; Ipo1; IPOA; SOD; Sod-1; SOD1; SODC
UniProt ID: (Human) Q16839, (Mouse) P08228, (Rat) P07632
Entrez Gene ID: (Human) 6647, (Mouse) 20655, (Rat) 24786
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.