Invitrogen
Manufacturer Code:PA552079
Catalog # PIPA552079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SOCS7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NAP-4; Nck, Ash and phospholipase C binding protein; Nck, Ash and phospholipase C gamma-binding protein; NCK-associated protein 4; SOCS-7; SOCS7 suppressor of cytokine signaling 7 NAP4 Nck Ash and phospholipase C binding protein; Suppressor of cytokine signaling 7
Gene Aliases: NAP4; NCKAP4; SOCS6; SOCS7
UniProt ID: (Human) O14512
Entrez Gene ID: (Human) 30837
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.