OriGene
Manufacturer Code:TA503054S
Catalog # 50-167-4027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI15F9 |
Immunogen | Full length human recombit protein of human SOCS3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATOD4 CIS3 Cish3 SOCS-3 SSI-3 SSI3; CIS-3; Cytokine-inducible SH2 protein 3; SOCS-3; SSI-3; STAT-induced STAT inhibitor 3; Suppressor of cytokine signaling 3
Gene Aliases: ATOD4; CIS3; Cish3; SOCS-3; SOCS3; SSI-3; SSI3
UniProt ID: (Human) O14543
Entrez Gene ID: (Human) 9021
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.