Invitrogen
Manufacturer Code:PA129534
Catalog # PIPA129534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (QLPGPIREFLDQYDAPL)corresponding to amino acids 209-225 of thehuman / rat / mouse SOCS 3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATOD4 CIS3 Cish3 MGC71791 SOCS-3 SSI-3 SSI3; CIS-3; cytokine inducible SH2-containing protein 3; Cytokine-inducible SH2 protein 3; E2a-Pbx1 target gene in fibroblasts 10; Protein EF-10; SOCS-3; SSI-3; STAT-induced STAT inhibitor 3; Suppressor of cytokine signaling 3
Gene Aliases: ATOD4; CIS3; Cish3; EF-10; Ef10; SOCS-3; SOCS3; SSI-3; SSI3
UniProt ID: (Human) O14543, (Mouse) O35718, (Rat) O88583
Entrez Gene ID: (Human) 9021, (Mouse) 12702, (Rat) 89829
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.