Abnova
Manufacturer Code:PAB0214
Catalog # 89-114-037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human SOCS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SOCS1.
For Research Use Only
Protein Aliases: CIS1; CISH1; cytokine-inducible SH2 protein 1; JAB; JAK binding protein; JAK-binding protein; SOCS-1; SSI-1; SSI1; STAT induced SH3 protein 1; STAT-induced STAT inhibitor 1; Suppressor of cytokine signaling 1; Tec-interacting protein 3; TIP-3; TIP3
Gene Aliases: CIS1; CISH1; JAB; SOCS-1; SOCS1; SSI-1; SSI1; TIP3
UniProt ID: (Human) O15524
Entrez Gene ID: (Human) 8651
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.