Novus Biologicals
Manufacturer Code:NBP274276C
Catalog # NBP274276C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D3 |
Immunogen | Full length recombinant protein of human SOCS-3 (NP_003946) produced in HEK293T cells. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CIS-3; CIS3CIS-3 Cish3 cytokine-induced SH2 protein 3 Cytokine-inducible SH2 protein 3 SOCS-3STAT-induced STAT inhibitor 3 SSI-3ATOD4 SSI3MGC71791 suppressor of cytokine signaling 3; Cytokine-inducible SH2 protein 3; SOCS-3; SSI-3; STAT-induced STAT inhibitor 3; Suppressor of cytokine signaling 3
Gene Aliases: ATOD4; CIS3; Cish3; SOCS-3; SOCS3; SSI-3; SSI3
UniProt ID: (Human) O14543
Entrez Gene ID: (Human) 9021
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.