Novus Biologicals
Manufacturer Code:NBP298892F
Catalog # NB106593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SOCS-1. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytokine-inducible SH2 protein 1; JAB; JABTec-interacting protein 3 JAK binding protein JAK-binding protein SOCS-1CIS1 SSI-1CISH1 SSI1cytokine-inducible SH2 protein 1 STAT induced SH3 protein 1 STAT-induced STAT inhibitor 1 suppressor of cytokine signaling 1 TIP-3 TIP3Cish1; JAK binding protein; JAK-binding protein; SOCS-1; SSI-1; STAT induced SH3 protein 1; STAT-induced STAT inhibitor 1; Suppressor of cytokine signaling 1; Tec-interacting protein 3; TIP-3
Gene Aliases: CIS1; CISH1; JAB; SOCS-1; SOCS1; SSI-1; SSI1; TIP3
UniProt ID: (Human) O15524
Entrez Gene ID: (Human) 8651
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.