OriGene
Manufacturer Code:CF800775
Catalog # 50-166-5310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-120 of human SOAT2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACACT2 ACAT2 ARGP2; ACAT-2; acyl Co-A: cholesterol acyltransferase 2; acyl coenzyme A:cholesterol acyltransferase 2; acyl-CoA:cholesterol acyltransferase 2; Acyl-coenzyme A:cholesterol acyltransferase 2; Cholesterol acyltransferase 2; Sterol O-acyltransferase 2
Gene Aliases: ACACT2; ACAT2; ARGP2; SOAT2
UniProt ID: (Human) O75908
Entrez Gene ID: (Human) 8435
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.