OriGene
Manufacturer Code:CF501250
Catalog # 50-166-0275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E2 |
Immunogen | Full length human recombit protein of human SNX9 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Protein SDP1; SDP1 SH3PX1 SH3PXD3A WISP; SH3 and PX domain-containing protein 1; SH3 and PX domain-containing protein 3A; Sorting nexin-9; Wiskott-Aldrich syndrome protein (WASP) interactor protein
Gene Aliases: SDP1; SH3PX1; SH3PXD3A; SNX9; WISP
UniProt ID: (Human) Q9Y5X1
Entrez Gene ID: (Human) 51429, (Dog) 476254
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.