Invitrogen
Manufacturer Code:MA537538
Catalog # PIMA537538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 335CT6.4.6 |
Immunogen | SNX6 recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2010006G21Rik 2610032J07Rik 2810425K19Rik AU018928 C85963 MSTP010 SNX6 sorting nexin 6 sorting nexin-6 Sorting nexin-6 N-terminally processed TFAF2 TRAF4-associated factor 2 tumor necrosis factor receptor-associated factor 4(TRAF4)-associated factor 2; Sorting nexin-6; Sorting nexin-6, N-terminally processed; TRAF4-associated factor 2; tumor necrosis factor receptor-associated factor 4(TRAF4)-associated factor 2
Gene Aliases: MSTP010; SNX6; TFAF2
UniProt ID: (Human) Q9UNH7
Entrez Gene ID: (Human) 58533
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.