OriGene
Manufacturer Code:CF811827
Catalog # 50-167-0865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 146-450 of human SNX4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATG24B; Sorting nexin-4
Gene Aliases: 1810036H14Rik; AI037066; ATG24B; SNX4
UniProt ID: (Human) O95219, (Mouse) Q91YJ2
Entrez Gene ID: (Human) 8723, (Mouse) 69150, (Rat) 360725
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.