Novus Biologicals
Manufacturer Code:NBP224514
Catalog # NBP224514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 120-170 of human Snx2 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTB-36H16.2; MGC5204 sorting nexin 2 sorting nexin-2 Transformation-related gene 9 protein TRG-9; Sorting nexin-2; Transformation-related gene 9 protein; TRG-9
Gene Aliases: SNX2; TRG-9; TRG9
UniProt ID: (Human) O60749
Entrez Gene ID: (Human) 6643
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.