Proteintech
Manufacturer Code:102751AP150UL
Catalog # 50-172-6171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SNX17 Fusion Protein Ag0355 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0064 SNX17 sorting nexin 17; Sorting nexin-17
Gene Aliases: 5830447M19Rik; AI790646; b2b1625.1Clo; D5Ertd260e; KIAA0064; mKIAA0064; SNX17
UniProt ID: (Human) Q15036, (Mouse) Q8BVL3, (Rat) Q6AYS6
Entrez Gene ID: (Human) 9784, (Mouse) 266781, (Rat) 298836
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.