Novus Biologicals
Manufacturer Code:NBP274263AF700
Catalog # N274263A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A4 |
Immunogen | Full length recombinant protein of human SNX12 (NP_037478) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: B-cell CLL/lymphoma 7 protein family member C B-cell CLL/lymphoma 7C; Sorting nexin-12
Gene Aliases: SNX12
UniProt ID: (Human) Q9UMY4
Entrez Gene ID: (Human) 29934
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.