Proteintech
Manufacturer Code:149531AP150UL
Catalog # 50-172-9861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SNX11 Fusion Protein Ag6791 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SNX11 sorting nexin 11; Sorting nexin-11
Gene Aliases: 4933439F10Rik; A930041K09Rik; AI117694; SNX11
UniProt ID: (Human) Q9Y5W9, (Mouse) Q91WL6
Entrez Gene ID: (Human) 29916, (Mouse) 74479, (Rat) 303493
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.