Invitrogen
Manufacturer Code:PA570054
Catalog # PIPA570054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SNX10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OPTB8 sorting nexin-10; Sorting nexin-10; Sorting nexin-10-like protein
Gene Aliases: 2410004M09Rik; OPTB8; SNX10
UniProt ID: (Human) Q9Y5X0, (Bovine) Q0IIL5, (Mouse) Q9CWT3
Entrez Gene ID: (Rabbit) 100356190, (Human) 29887, (Pig) 100520876, (Horse) 100068610, (Dog) 475261, (Bovine) 508836, (Rat) 297096, (Mouse) 71982, (Guinea pig) 100714844
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.