Proteintech
Manufacturer Code:103041AP150UL
Catalog # 50-172-6193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SNX1 Fusion Protein Ag0499 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsT17379 SNX1 SNX1A sorting nexin 1 Vps5; sorting nexin 1A; Sorting nexin-1
Gene Aliases: HsT17379; SNX1; VPS5
UniProt ID: (Human) Q13596, (Mouse) Q9WV80, (Rat) Q99N27
Entrez Gene ID: (Human) 6642, (Mouse) 56440, (Rat) 84471
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.