Invitrogen
Manufacturer Code:PA529070
Catalog # PIPA529070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 239 and 516 of Human NCOA62 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bx42 NCOA-62 PRPF45 Prp45 SKIIP SKIP SKI interacting protein SKI-interacting protein SNW domain-containing protein 1 homolog of Drosophila BX42 nuclear protein SkiP nuclear receptor coactivator NCoA-62 nuclear receptor coactivator 62-kD; homolog of Drosophila BX42; Nuclear protein SkiP; Nuclear receptor coactivator NCoA-62; nuclear receptor coactivator, 62-kD; SKI interacting protein; SKI-interacting protein; SNW domain-containing protein 1
Gene Aliases: 2310008B08Rik; AW048543; Bx42; NCOA-62; Prp45; PRPF45; SKIIP; SKIP; SNW1
UniProt ID: (Human) Q13573, (Mouse) Q9CSN1
Entrez Gene ID: (Human) 22938, (Mouse) 66354
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.