Novus Biologicals
Manufacturer Code:NB1002367
Catalog # NB1002367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human Nuclear Receptor Coactivator 62 kD (SKIIP SKI Interacting Protein) using the numbering given in entry NP_036377.1 (GeneID 22938). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bx42 homolog of Drosophila BX42 MGC119379 NCOA-62 Nuclear protein SkiP Nuclear receptor coactivator NCoA-62 nuclear receptor coactivator 62-kD Prp45 PRPF45 SKI interacting protein Ski-interacting protein SKIPSKIIP SNW domain containing 1 SNW domain-containing protein 1; homolog of Drosophila BX42; Nuclear protein SkiP; Nuclear receptor coactivator NCoA-62; nuclear receptor coactivator, 62-kD; SKI interacting protein; SKI-interacting protein; SNW domain-containing protein 1
Gene Aliases: Bx42; NCOA-62; Prp45; PRPF45; SKIIP; SKIP; SNW1
UniProt ID: (Human) Q13573
Entrez Gene ID: (Human) 22938
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.