Abnova
Manufacturer Code:PAB12847
Catalog # 89-116-127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 16 amino acids of human SNW1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SNW1.
For Research Use Only
Protein Aliases: Bx42; homolog of Drosophila BX42; MGC119379; NCOA-62; Nuclear protein SkiP; Nuclear receptor coactivator NCoA-62; nuclear receptor coactivator, 62-kD; Prp45; PRPF45; SKI interacting protein; SKI-interacting protein; SKIIP; SKIP; SNW domain-containing protein 1
Gene Aliases: Bx42; NCOA-62; Prp45; PRPF45; SKIIP; SKIP; SNW1
UniProt ID: (Human) Q13573
Entrez Gene ID: (Human) 22938
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.