Invitrogen
Manufacturer Code:PA569085
Catalog # PIPA569085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SNUPN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KPNBL RNUT1 Snurportin1 RNA U transporter 1 RNA U transporter 1 snurportin-1; RNA U transporter 1; RNA, U transporter 1; Snurportin-1; snurportin-1-like protein
Gene Aliases: 0610031A09Rik; KPNBL; RNUT1; SNUPN; Snupn1; Snurportin1; SPN1; wu:fc41a03; zgc:77819
UniProt ID: (Bovine) Q2TBK8, (Human) O95149, (Rat) Q68FP5, (Mouse) Q80W37
Entrez Gene ID: (Rabbit) 100343101, (Bovine) 515588, (Human) 10073, (Dog) 487655, (Horse) 100051100, (Rat) 316108, (Mouse) 66069, (Guinea pig) 100734874, (Zebrafish) 324743
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.