OriGene
Manufacturer Code:CF502040
Catalog # 50-166-0980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E12 |
Immunogen | Full length human recombit protein of human SNTA1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 59 kDa dystrophin-associated protein A1 acidic component 1; acidic alpha 1 syntrophin; Alpha-1-syntrophin; dystrophin-associated protein A1, 59kDa, acidic component; LQT12 SNT1 TACIP1 dJ1187J4.5; Pro-TGF-alpha cytoplasmic domain-interacting protein 1; syntrophin, acidic 1; syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component); Syntrophin-1; TACIP1
Gene Aliases: dJ1187J4.5; LQT12; SNT1; SNTA1; TACIP1
UniProt ID: (Human) Q13424
Entrez Gene ID: (Human) 6640, (Dog) 485837, (Rat) 362242
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.