Invitrogen
Manufacturer Code:PA5113696
Catalog # PIPA5113696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SNRPF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010013O18Rik BOS_5517 LOC100348389 PBSCF Sm protein F Small nuclear ribonucleoprotein F small nuclear ribonucleoprotein polypeptide F SmF Sm-F snRNP-F SNRPF SNRPF protein-like snrpfl; Sm protein F; Sm-F; Small nuclear ribonucleoprotein F; snRNP-F
Gene Aliases: PBSCF; Sm-F; SMF; snRNP-F; SNRPF
UniProt ID: (Human) P62306
Entrez Gene ID: (Human) 6636
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.