Novus Biologicals
Manufacturer Code:NBP157464
Catalog # NBP157464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SNRPF(small nuclear ribonucleoprotein polypeptide F) The peptide sequence was selected from the middle region of SNRPF. Peptide sequence GYMNMQLANTEEYIDGALSGHLGEVLIRCNNVLYIRGVEEEEEDGEMRE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PBSCF Sm protein F small nuclear ribonucleoprotein polypeptide F SMF Sm-Fsmall nuclear ribonucleoprotein F snRNP-F; Sm protein F; Sm-F; Small nuclear ribonucleoprotein F; snRNP-F
Gene Aliases: PBSCF; Sm-F; SMF; snRNP-F; SNRPF
UniProt ID: (Human) P62306
Entrez Gene ID: (Human) 6636
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.