Proteintech
Manufacturer Code:149771AP150UL
Catalog # 50-172-9880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SNRPF Fusion Protein Ag6889 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PBSCF Sm F Sm protein F SMF snRNP F SNRPF; Sm protein F; Sm-F; Small nuclear ribonucleoprotein F; snRNP-F
Gene Aliases: 2010013O18Rik; PBSCF; Sm-F; SMF; snRNP-F; SNRPF
UniProt ID: (Human) P62306, (Mouse) P62307
Entrez Gene ID: (Human) 6636, (Mouse) 69878, (Rat) 680737
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.