Invitrogen
Manufacturer Code:PA576746
Catalog # PIPA576746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SNRPE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B rafB-rafBrafC76690HYPT11RUXE&lowbarHUMANSm protein ESm-ESmall nuclear ribonucleoprotein ESmEsnRNP-EsnRNPEsnrpe; Sm protein E; Sm-E; Small nuclear ribonucleoprotein E; snRNP-E
Gene Aliases: AL022645; C76690; HYPT11; Sm-E; SME; snRNP-E; SNRPE
UniProt ID: (Human) B2R5B9, (Mouse) P62305
Entrez Gene ID: (Human) 6635, (Mouse) 20643
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.