Invitrogen
Manufacturer Code:PA570618
Catalog # PIPA570618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SNRPE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sm protein E; Sm-E; Small nuclear ribonucleoprotein E; smE; snRNP-E; SNRPE
Gene Aliases: AL022645; C76690; HYPT11; Sm-E; SME; snRNP-E; SNRPE
UniProt ID: (Bovine) A4FUI2, (Human) B2R5B9, (Mouse) P62305
Entrez Gene ID: (Dog) 100855941, (Bovine) 531493, (Human) 6635, (Goat) 100861346, (Guinea pig) 100724868, (Rat) 100360682, (Mouse) 20643
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.