Novus Biologicals
Manufacturer Code:NBP243792
Catalog # NBP243792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human SNRPE. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-raf Sm protein E small nuclear ribonucleoprotein E small nuclear ribonucleoprotein polypeptide E SmE Sm-ESME snRNP-E; Sm protein E; Sm-E; Small nuclear ribonucleoprotein E; snRNP-E
Gene Aliases: HYPT11; Sm-E; SME; snRNP-E; SNRPE
UniProt ID: (Human) B2R5B9
Entrez Gene ID: (Human) 6635
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.