Invitrogen
Manufacturer Code:PA551524
Catalog # PIPA551524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SNRPD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sm-D3; small nuclear ribonucleoprotein D3; small nuclear ribonucleoprotein D3 polypeptide 18kDa; Small nuclear ribonucleoprotein Sm D3; SMD3 Sm-D3 small nuclear ribonucleoprotein Sm D3 small nuclear ribonucleoprotein polypeptide D3 snRNP core protein D3; snRNP core protein D3
Gene Aliases: 1700043E15Rik; 2310009E13Rik; AW046420; Sm-D3; SMD3; SNRPD3
UniProt ID: (Human) P62318, (Mouse) P62320
Entrez Gene ID: (Human) 6634, (Rat) 687711, (Mouse) 67332
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.