Novus Biologicals
Manufacturer Code:NBP222305
Catalog # NBP222305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1 to 50 of human Small Nuclear Ribonucleoprotein D3 Polypeptide 18kDa using the numbering given in entry NP_004166.1 (GeneID 6634). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sm-D3; small nuclear ribonucleoprotein D3 polypeptide 18kDa; small nuclear ribonucleoprotein D3 polypeptide 18kDa small nuclear ribonucleoprotein Sm D3 snRNP core protein D3; Small nuclear ribonucleoprotein Sm D3; snRNP core protein D3
Gene Aliases: Sm-D3; SMD3; SNRPD3
UniProt ID: (Human) P62318
Entrez Gene ID: (Human) 6634
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.