Novus Biologicals
Manufacturer Code:NBP29370420UL
Catalog # NB029272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 8-121 of human SNRPD3 (NP_001265585). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sm-D3; small nuclear ribonucleoprotein D3 polypeptide 18kDa; small nuclear ribonucleoprotein D3 polypeptide 18kDa small nuclear ribonucleoprotein Sm D3 snRNP core protein D3; Small nuclear ribonucleoprotein Sm D3; snRNP core protein D3
Gene Aliases: Sm-D3; SMD3; SNRPD3
UniProt ID: (Human) P62318
Entrez Gene ID: (Human) 6634
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.