Invitrogen
Manufacturer Code:PA527547
Catalog # PIPA527547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 101 of Human SNRPD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sm-D2; small nuclear ribonucleoprotein D2 polypeptide 16.5kDa; Small nuclear ribonucleoprotein Sm D2; SMD2 SNRPD1 Sm-D2 small nuclear ribonucleoprotein Sm D2 snRNP core protein D2; snRNP core protein D2
Gene Aliases: 1810009A06Rik; Sm-D2; SMD2; SNRPD1; SNRPD2
UniProt ID: (Human) P62316, (Mouse) P62317
Entrez Gene ID: (Human) 6633, (Mouse) 107686
Molecular Function:
RNA binding protein
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.