Novus Biologicals
Manufacturer Code:NBP222299
Catalog # NBP222299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 109 to 159 of human Small Nuclear Ribonucleoprotein Polypeptide C using the numbering given in entry NP_003084.1 (GeneID 6631). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ20302 small nuclear ribonucleoprotein polypeptide C U1 small nuclear ribonucleoprotein C U1 small nuclear RNP specific C U1 snRNP C U1 snRNP protein C U1C U1-C Yhc1; SNRPC; U1 small nuclear ribonucleoprotein C; U1 small nuclear RNP specific C; U1 snRNP C; U1 snRNP protein C; U1-C; U1C
Gene Aliases: SNRPC; U1C; Yhc1
UniProt ID: (Human) P09234
Entrez Gene ID: (Human) 6631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.