OriGene
Manufacturer Code:TA808092
Catalog # 50-168-1907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D9 |
Immunogen | Full length human recombit protein of human SNRPB2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Msl1 U2B&prime′ small nuclear ribonucleoprotein polypeptide B; U2 small nuclear ribonucleoprotein B''; U2 snRNP B''
Gene Aliases: Msl1; SNRPB2; U2B''
UniProt ID: (Human) P08579
Entrez Gene ID: (Human) 6629
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.