Invitrogen
Manufacturer Code:PA584991
Catalog # PIPA584991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SNRPB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Msl1 U2B&prime′ small nuclear ribonucleoprotein polypeptide B; U2 small nuclear ribonucleoprotein B''; U2 snRNP B''
Gene Aliases: Msl1; SNRPB2; U2B''
UniProt ID: (Human) P08579
Entrez Gene ID: (Human) 6629
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.