Invitrogen
Manufacturer Code:MA513449
Catalog # PIMA513449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Y12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B polypeptide of Sm protein; Sm protein B; Sm protein B/B'; Sm-B; Sm-B/B'; sm-B/Sm-B'; SM11; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; Small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; smB; snRNP-B
Gene Aliases: AL024368; AU018828; CCMS; COD; SM-B; Sm-B/B'; SM11; SMB; SmB/B'; SmB/SmB'; SNRNP-B; SNRPB; SNRPB1
UniProt ID: (Human) Q9UIS5, (Mouse) P27048, (Rat) P17136
Entrez Gene ID: (Human) 6628, (Rabbit) 100341600, (Mouse) 20638, (Rat) 171365
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.