Invitrogen
Manufacturer Code:PA5110407
Catalog # PIPA5110407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SNRPB (NP_003082.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AL024368 AU018828 CCMS COD Sm protein B Sm protein B/B&prime SM11 Small nuclear ribonucleoprotein-associated protein B Small nuclear ribonucleoprotein-associated proteins B and B&prime SMB Sm-B Sm-B/B&prime SmB/B&prime SmB/SmB&prime snRNP-B Snrpb SNRPB1; B polypeptide of Sm protein; Sm protein B; Sm protein B/B'; Sm-B; Sm-B/B'; sm-B/Sm-B'; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; Small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; snRNP-B
Gene Aliases: AL024368; AU018828; CCMS; COD; SM-B; Sm-B/B'; SM11; SMB; SmB/B'; SmB/SmB'; SNRNP-B; SNRPB; SNRPB1
UniProt ID: (Human) Q9UIS5, (Mouse) P27048
Entrez Gene ID: (Human) 6628, (Mouse) 20638
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.