Invitrogen
Manufacturer Code:PA541728
Catalog # PIPA541728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SNRP70 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: small nuclear ribonucleoprotein 70kDa; small nuclear ribonucleoprotein 70kDa (U1); Small nuclear ribonucleoprotein 70kDa (U1) RPU1 Snp1 U1AP U170K U1RNP RNPU1Z SNRP70 U1-70K; small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen); small nuclear ribonucleoprotein, U1 70kDa subunit; U1 small nuclear ribonucleoprotein 70 kDa; U1 small nuclear ribonucleoprotein polypeptide A; U1 small ribonucleoprotein 70kd; U1 snRNP 70 kDa; U1-70K
Gene Aliases: 2700022N21Rik; 3200002N22Rik; AI325098; R74807; RNPU1Z; Rnu1p70; Rnulp70; RPU1; Snp1; SNRNP70; SNRP70; Srnp70; U1-70; U1-70K; U170K; U1AP; U1AP1; U1RNP; wu:fc20b04; zgc:136450
UniProt ID: (Bovine) Q1RMR2, (Human) P08621, (Mouse) Q62376
Entrez Gene ID: (Dog) 484388, (Bovine) 535113, (Human) 6625, (Rabbit) 100354916, (Horse) 100054907, (Mouse) 20637, (Guinea pig) 100729616, (Rat) 361574, (Zebrafish) 445398
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.