Invitrogen
Manufacturer Code:PA5115943
Catalog # PIPA5115943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SNRP70 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700022N21Rik 3200002N22Rik AI325098 R74807 RNPU1Z Rnu1p70 Rnulp70 RPU1 small nuclear ribonucleoprotein 70 (U1) small nuclear ribonucleoprotein 70kDa (U1) small nuclear ribonucleoprotein U1 subunit 70 small nuclear ribonucleoprotein U1 70kDa subunit Snp1 Snrnp70 SNRP70 Srnp70 U1 small nuclear ribonucleoprotein 70 kDa U1 small nuclear ribonucleoprotein polypeptide A U1 snRNP 70 kDa U1-70 U170K U1-70K U1AP U1AP1 U1RNP; small nuclear ribonucleoprotein 70kDa (U1); small nuclear ribonucleoprotein, U1 70kDa subunit; U1 small nuclear ribonucleoprotein 70 kDa; U1 small nuclear ribonucleoprotein polypeptide A; U1 snRNP 70 kDa
Gene Aliases: 2700022N21Rik; 3200002N22Rik; AI325098; R74807; RNPU1Z; Rnu1p70; Rnulp70; RPU1; Snp1; SNRNP70; SNRP70; Srnp70; U1-70; U1-70K; U170K; U1AP; U1AP1; U1RNP
UniProt ID: (Human) P08621, (Mouse) Q62376
Entrez Gene ID: (Human) 6625, (Mouse) 20637, (Rat) 361574
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.