Invitrogen
Manufacturer Code:PA5113695
Catalog # PIPA5113695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal region of human SNRP70 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700022N21Rik 3200002N22Rik AI325098 BOS_17698 R74807 RNPU1Z Rnu1p70 Rnulp70 RPU1 small nuclear ribonucleoprotein 70 (U1) small nuclear ribonucleoprotein 70kDa small nuclear ribonucleoprotein 70kDa (U1) small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen) small nuclear ribonucleoprotein U1 subunit 70 small nuclear ribonucleoprotein U1 70kDa subunit Snp1 Snrnp70 SNRP70 snrp70 protein Srnp70 U1 small nuclear ribonucleoprotein 70 kDa U1 small nuclear ribonucleoprotein polypeptide A U1 small ribonucleoprotein 70kd U1 snRNP 70 kDa U1-70 U170K U1-70K U1AP U1AP1 U1RNP wu:fc20b04 zgc:136450; small nuclear ribonucleoprotein 70kDa (U1); small nuclear ribonucleoprotein, U1 70kDa subunit; U1 small nuclear ribonucleoprotein 70 kDa; U1 snRNP 70 kDa
Gene Aliases: RNPU1Z; RPU1; Snp1; SNRNP70; SNRP70; U1-70K; U170K; U1AP; U1AP1; U1RNP
UniProt ID: (Human) P08621
Entrez Gene ID: (Human) 6625
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.