Invitrogen
Manufacturer Code:PA5110406
Catalog # PIPA5110406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SNRNP70 (NP_003080.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700022N21Rik 3200002N22Rik AI325098 BOS_17698 R74807 RNPU1Z Rnu1p70 Rnulp70 RPU1 Snp1 Snrnp70 Snrp70 Srnp70 U1 small nuclear ribonucleoprotein 70 kDa U1 snRNP 70 kDa U1-70 U170K U1-70K U1AP U1AP1 U1RNP wu:fc20b04 zgc:136450; small nuclear ribonucleoprotein 70kDa (U1); small nuclear ribonucleoprotein, U1 70kDa subunit; U1 small nuclear ribonucleoprotein 70 kDa; U1 small nuclear ribonucleoprotein polypeptide A; U1 snRNP 70 kDa
Gene Aliases: 2700022N21Rik; 3200002N22Rik; AI325098; R74807; RNPU1Z; Rnu1p70; Rnulp70; RPU1; Snp1; SNRNP70; SNRP70; Srnp70; U1-70; U1-70K; U170K; U1AP; U1AP1; U1RNP
UniProt ID: (Human) P08621, (Mouse) Q62376
Entrez Gene ID: (Human) 6625, (Mouse) 20637, (Rat) 361574
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.