Novus Biologicals
Manufacturer Code:NBP232449
Catalog # NBP232449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: WSEMLGVGDFYYELGVQIIEVCLALKHRNGGLITLEELHQQVLKGRGKFAQDVSQDDLIRAIKKLKALGTGFGIIPVGGTYL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAP30 subunit of ELL complex; EAP30 subunit of ELL complex EAP30hVps22 ELL-associated protein of 30 kDa ESCRT-II complex subunit VPS22 SNF8 ESCRT-II complex subunit homolog (S. cerevisiae) vacuolar-sorting protein SNF8 VPS22Dot3; ELL-associated protein of 30 kDa; ESCRT-II complex subunit VPS22; hVps22; SNF8, ESCRT-II complex subunit, homolog; Vacuolar-sorting protein SNF8
Gene Aliases: Dot3; EAP30; SNF8; VPS22
UniProt ID: (Human) Q96H20
Entrez Gene ID: (Human) 11267
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.