Novus Biologicals
Manufacturer Code:NB10057525
Catalog # NB10057525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human SMARCB1/SNF5 using the numbering given in entry NP_003064.2 (GeneID 6598). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAF47; BAF47 BAF47hSNF5 BRG1-associated factor 47 hSNFS Ini1 INI1RTPS1 Integrase interactor 1 protein malignant rhabdoid tumor suppressor RDTSNF5 homolog Sfh1p SNF5 SNF5L1 Snr1 subfamily b member 1 sucrose nonfermenting yeast homolog-like 1 SWI/SNF related matrix associated actin dependent regulator of chromatin SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily B member 1; BRG1-associated factor 47; hSNF5; Integrase interactor 1 protein; malignant rhabdoid tumor suppressor; protein phosphatase 1, regulatory subunit 144; SNF5 homolog; sucrose nonfermenting, yeast, homolog-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1; SWI/SNF-related matrix-associated protein
Gene Aliases: BAF47; CSS3; hSNFS; INI1; MRD15; PPP1R144; RDT; RTPS1; Sfh1p; SMARCB1; SNF5; SNF5L1; Snr1; SWNTS1
UniProt ID: (Human) Q76N08
Entrez Gene ID: (Human) 6598
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.