Proteintech
Manufacturer Code:602651IG150UL
Catalog # 50-173-6362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A6A4 |
Immunogen | SND1 Fusion Protein Ag1200 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa coactivator; 100 kDa coactivator EBNA2 coactivator p100 p100 p100 co activator SND1 TDRD11; EBNA-2 co-activator (100kD); EBNA2 coactivator p100; p100 co-activator; p100 EBNA2 co-activator; SND1-BRAF fusion; staphylococcal nuclease domain containing 1; Staphylococcal nuclease domain-containing protein 1; testis tissue sperm-binding protein Li 82P; Tudor domain-containing protein 11; Tudor-SN
Gene Aliases: AL033314; p100; SND1; TDRD11; Tudor-SN
UniProt ID: (Human) Q7KZF4, (Mouse) Q78PY7
Entrez Gene ID: (Human) 27044, (Mouse) 56463
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.