Invitrogen
Manufacturer Code:PA540562
Catalog # PIPA540562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SND1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa coactivator; 4SNc-Tudor domain protein; EBNA-2 co-activator (100kD); EBNA2 coactivator p100; p100 co-activator; p100 EBNA2 co-activator; p100L; p100S; similar to Homo sapiens EBNA-2 co-activator (100kDa coactivator); p105 coactivator; SND p102; SND1-BRAF fusion; Staphylococcal nuclease and tudor domain containing 1 p100 TDRD11; staphylococcal nuclease domain containing 1; Staphylococcal nuclease domain-containing protein 1; testis tissue sperm-binding protein Li 82P; Tudor domain-containing protein 11; Tudor-SN
Gene Aliases: AL033314; p100; SN4TDR; SND p102; SND1; TDRD11; Tudor-SN; wu:fc27g10
UniProt ID: (Human) Q7KZF4, (Bovine) Q863B3, (Rat) Q66X93, (Mouse) Q78PY7, (Zebrafish) Q7ZT42
Entrez Gene ID: (Human) 27044, (Bovine) 404098, (Rabbit) 100340569, (Horse) 100056692, (Dog) 475204, (Rat) 64635, (Mouse) 56463, (Guinea pig) 100726680, (Zebrafish) 324404
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.