Proteintech
Manufacturer Code:104981AP150UL
Catalog # 50-172-6353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SNCB Fusion Protein Ag0772 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta synuclein SNCB synuclein beta; Beta-synuclein; Phosphoneuroprotein 14; PNP 14
Gene Aliases: AI838531; betaSYN; SNCB
UniProt ID: (Human) Q16143, (Mouse) Q91ZZ3, (Rat) Q63754
Entrez Gene ID: (Human) 6620, (Mouse) 104069, (Rat) 113893
Molecular Function:
chaperone
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.