OriGene
Manufacturer Code:CF506533
Catalog # 50-166-4336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F6 |
Immunogen | Full length human recombit protein of human SNCA produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-synuclein; NACP; NACP PARK1 PARK4 PD1; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; Non-A4 component of amyloid precursor; synuclein alpha-140; synuclein, alpha (non A4 component of amyloid precursor)
Gene Aliases: NACP; PARK1; PARK4; PD1; SNCA
UniProt ID: (Human) P37840
Entrez Gene ID: (Human) 6622
Molecular Function:
chaperone
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.