Abnova
Manufacturer Code:PAB0770
Catalog # 89-114-410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 108-120 of human SNCA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Sheep polyclonal antibody raised against synthetic peptide of SNCA.
For Research Use Only
Protein Aliases: Alpha-synuclein; MGC110988; NACP; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; synuclein alpha-140; synuclein, alpha (non A4 component of amyloid precursor)
Gene Aliases: NACP; PARK1; PARK4; PD1; SNCA
UniProt ID: (Human) P37840
Entrez Gene ID: (Human) 6622
Molecular Function: chaperone membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.