Invitrogen
Manufacturer Code:PA546857
Catalog # PIPA546857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SNAPC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Proximal sequence element-binding transcription factor subunit gamma; PSE-binding factor subunit gamma; PTF subunit gamma; Small nuclear RNA activating complex SNAP43 PTFgamma; small nuclear RNA activating complex, polypeptide 1, 43kD; small nuclear RNA activating complex, polypeptide 1, 43kDa; Small nuclear RNA-activating complex polypeptide 1; SNAPc 43 kDa subunit; SNAPc subunit 1; snRNA-activating protein complex 43 kDa subunit; snRNA-activating protein complex subunit 1
Gene Aliases: PTFgamma; SNAP43; SNAPC1
UniProt ID: (Human) Q16533
Entrez Gene ID: (Human) 6617
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.