Proteintech
Manufacturer Code:149031AP150UL
Catalog # 50-172-9821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SNAP25 Fusion Protein Ag6695 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: blind drunk; resistance to inhibitors of cholinesterase 4 homolog; RIC 4 RIC4 SEC9 SNAP SNAP 25 SNAP25 SUP Super protein; SNAP-25; SUP; Super protein; Synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25kDa
Gene Aliases: bA416N4.2; Bdr; CMS18; dJ1068F16.2; GENA70; RIC-4; RIC4; SEC9; SNAP; SNAP-25; SNAP-25a; SNAP-25B; SNAP25; sp; SUP
UniProt ID: (Human) P60880, (Rat) P60881, (Mouse) P60879
Entrez Gene ID: (Human) 6616, (Rat) 25012, (Mouse) 20614
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.